siRNA Details
| Gene Symbol: LUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002345.3 | NP_002336.1 | 2 | 628 | ||
| GenBank mRNA | AK223177.1 | - | 2 | 313 | ||
| AK312682.1 | - | 2 | 331 | |||
| BC007038.1 | - | 2 | 380 | 3950745 | ||
| BC035997.1 | - | 2 | 302 | 4688606 | ||
| BG548387.1 | - | 2 | 698 | |||
| BT006707.1 | - | 1 | 239 | |||
| U18728.1 | - | 2 | 319 | |||
| U21128.1 | - | 2 | 323 | |||

LUM
Genomic Map