siRNA Details
| Gene Symbol: Rnf181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025607.3 | NP_079883.3 | 1, 2 | 148 | ||
| GenBank mRNA | AK010854.1 | - | 1, 2 | 161 | ||
| AK159788.1 | - | 1, 2 | 161 | |||
| AK166904.1 | - | 1, 2 | 160 | |||
| AK166995.1 | - | 1, 2 | 161 | |||
| AK167396.1 | - | 1, 2 | 161 | |||
| AK167633.1 | - | 1, 2 | 161 | |||
| AK167935.1 | - | 1, 2 | 161 | |||
| AK168670.1 | - | 1, 2 | 162 | |||
| BC005559.1 | - | 1, 2 | 172 | |||
| BC083119.1 | - | 1, 2 | 149 | 5374213 | ||

Rnf181
Genomic Map