siRNA Details
| Gene Symbol: Cul7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025611.5 | NP_079887.3 | 11 | 2769 | ||
| GenBank mRNA | AK147156.1 | - | 11 | 2527 | ||
| AK147905.1 | - | 11 | 2488 | |||
| AK155583.1 | - | 11 | 2777 | |||
| AK170580.1 | - | 11 | 2773 | |||
| AK170808.1 | - | 11 | 2543 | |||
| AK172882.1 | - | 10 | 1842 | |||
| BC019645.1 | - | 1 | 57 | |||
| BC059865.1 | - | 8 | 1489 | |||
| BC065096.1 | - | 10 | 2256 | |||
| BC138998.1 | - | 11 | 2707 | 8862020 | ||
| BC139000.1 | - | 11 | 2707 | 8862022 | ||

Cul7
Genomic Map