siRNA Details
| Gene Symbol: Rwdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025614.3 | NP_079890.1 | 7 | 840 | ||
| GenBank mRNA | AF503942.1 | - | 7 | 732 | ||
| AK003262.2 | - | 7 | 794 | |||
| AK005278.1 | - | 7 | 752 | |||
| AK009510.1 | - | 7 | 709 | |||
| AK011277.1 | - | 7 | 840 | |||
| BC062136.1 | - | 7 | 722 | 6508909 | ||

Rwdd1
Genomic Map