siRNA Details
| Gene Symbol: LUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002345.3 | NP_002336.1 | 2 | 678 | ||
| GenBank mRNA | AK223177.1 | - | 2 | 363 | ||
| AK312682.1 | - | 2 | 381 | |||
| BC007038.1 | - | 2 | 430 | 3950745 | ||
| BC035997.1 | - | 2 | 352 | 4688606 | ||
| BT006707.1 | - | 1 | 289 | |||
| U18728.1 | - | 2 | 369 | |||
| U21128.1 | - | 2 | 373 | |||

LUM
Genomic Map