siRNA Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025616.3 | NP_079892.1 | 3 | 285 | ||
| GenBank mRNA | AK008340.1 | - | 3 | 286 | ||
| AK008382.1 | - | 3 | 283 | |||
| AK010832.1 | - | 3 | 251 | |||
| AK012977.1 | - | 3 | 282 | |||
| AK088074.1 | - | 3 | 278 | |||
| AK167473.1 | - | 3 | 288 | |||
| AK178698.1 | - | 2 | 59 | |||
| AY551342.1 | - | 3 | 271 | |||
| BC010303.1 | - | 3 | 260 | 3597676 | ||
| BC037056.1 | - | 3 | 287 | |||

Timm50
Genomic Map