siRNA Details
| Gene Symbol: Tceanc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025617.2 | NP_079893.1 | 3 | 359 | ||
| GenBank mRNA | AK008715.1 | - | 3 | 314 | ||
| AK011991.1 | - | 3 | 300 | |||
| BC028321.1 | - | 3 | 297 | 3992258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tceanc2| Gene Symbol: Tceanc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025617.2 | NP_079893.1 | 3 | 359 | ||
| GenBank mRNA | AK008715.1 | - | 3 | 314 | ||
| AK011991.1 | - | 3 | 300 | |||
| BC028321.1 | - | 3 | 297 | 3992258 | ||