siRNA Details
| Gene Symbol: Tceanc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025617.2 | NP_079893.1 | 2 | 214 | ||
| GenBank mRNA | AK008694.1 | - | 2 | 357 | ||
| AK008715.1 | - | 2 | 169 | |||
| AK011991.1 | - | 2 | 155 | |||
| BC028321.1 | - | 2 | 152 | 3992258 | ||

Tceanc2
Genomic Map