siRNA Details
| Gene Symbol: Ubxn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024432.2 | NP_077752.1 | 1 | 142 | ||
| GenBank mRNA | AF272895.1 | - | 1 | 157 | ||
| AK008942.1 | - | 1 | 152 | |||
| AK075705.1 | - | 1 | 171 | |||
| AK082510.1 | - | 181 | ||||
| AK148170.1 | - | 110 | ||||
| AK161273.1 | - | 1 | 163 | |||
| BC016412.1 | - | 1 | 71 | 4215883 | ||
| BC049963.1 | - | 1 | 142 | 2655935 | ||

Ubxn6
Genomic Map