siRNA Details
| Gene Symbol: Ubxn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024432.2 | NP_077752.1 | 6 | 628 | ||
| GenBank mRNA | AF272895.1 | - | 6 | 643 | ||
| AK008942.1 | - | 6 | 637 | |||
| AK075705.1 | - | 5 | 816 | |||
| AK082510.1 | - | 667 | ||||
| AK131187.1 | - | 1 | 1524 | |||
| AK148170.1 | - | 596 | ||||
| AK161273.1 | - | 6 | 649 | |||
| BC016412.1 | - | 6 | 557 | 4215883 | ||
| BC049963.1 | - | 6 | 628 | 2655935 | ||

Ubxn6
Genomic Map