siRNA Details
| Gene Symbol: 2310050C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025621.2 | NP_079897.2 | 2 | 521 | ||
| GenBank mRNA | AK003712.1 | - | 1 | 172 | ||
| AK009900.1 | - | 1 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310050C09Rik| Gene Symbol: 2310050C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025621.2 | NP_079897.2 | 2 | 521 | ||
| GenBank mRNA | AK003712.1 | - | 1 | 172 | ||
| AK009900.1 | - | 1 | 448 | |||