siRNA Details
Gene Symbol: 2310050C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025621.2 | NP_079897.2 | 2 | 416 | ||
GenBank mRNA | AK003712.1 | - | 1 | 67 | ||
AK009900.1 | - | 1 | 343 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310050C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025621.2 | NP_079897.2 | 2 | 416 | ||
GenBank mRNA | AK003712.1 | - | 1 | 67 | ||
AK009900.1 | - | 1 | 343 |