siRNA Details
| Gene Symbol: Nipsnap3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025623.2 | NP_079899.1 | 2 | 314 | ||
| GenBank mRNA | AF353623.1 | - | 2 | 303 | ||
| AK004458.1 | - | 2 | 313 | |||
| AK008786.1 | - | 2 | 310 | |||
| AK012480.1 | - | 2 | 314 | |||
| BC019505.1 | - | 2 | 299 | 4164310 | ||

Nipsnap3b
Genomic Map