siRNA Details
| Gene Symbol: Rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175109.3 | NP_780318.1 | 4 | 330 | ||
| GenBank mRNA | AB233218.1 | - | 4 | 326 | ||
| AK010954.1 | - | 4 | 314 | |||
| AK075974.1 | - | 4 | 311 | |||
| BC038691.1 | - | 4 | 316 | 1495043 | ||
| BC050115.1 | - | 4 | 319 | 30017673 | ||
| BC065381.1 | - | 3 | 263 | |||
| BY237968.1 | - | 4 | 330 | |||

Rps19bp1
Genomic Map