siRNA Details
Gene Symbol: Aggf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025630.3 | NP_079906.2 | 12 | 2044 | ||
GenBank mRNA | AK009533.1 | - | 8 | 1007 | ||
AK132575.1 | - | 12 | 2045 | |||
AY500995.1 | - | 12 | 2017 | |||
BC027286.1 | - | 7 | 720 | |||
BC052410.1 | - | 12 | 2021 | 5716440 |
