siRNA Details
| Gene Symbol: Aggf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025630.3 | NP_079906.2 | 7 | 1565 | ||
| GenBank mRNA | AK009533.1 | - | 3 | 528 | ||
| AK132575.1 | - | 7 | 1566 | |||
| AY500995.1 | - | 7 | 1538 | |||
| BC027286.1 | - | 2 | 241 | |||
| BC052410.1 | - | 7 | 1542 | 5716440 | ||

Aggf1
Genomic Map