siRNA Details
| Gene Symbol: Gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025638.2 | NP_079914.1 | 10 | 1049 | ||
| GenBank mRNA | AK011487.1 | - | 10 | 972 | ||
| AK013864.1 | - | 10 | 975 | |||
| AK039183.1 | - | 1 | 31 | |||
| AK167812.1 | - | 10 | 1032 | |||
| BC016541.1 | - | 10 | 972 | 3602449 | ||

Gdpd1
Genomic Map