siRNA Details
| Gene Symbol: Gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025638.2 | NP_079914.1 | 2, 3 | 348 | ||
| GenBank mRNA | AK011487.1 | - | 2, 3 | 271 | ||
| AK013864.1 | - | 2, 3 | 274 | |||
| AK016023.1 | - | 2, 3 | 274 | |||
| AK167812.1 | - | 2, 3 | 331 | |||
| BC016541.1 | - | 2, 3 | 271 | 3602449 | ||
| BY282485.1 | - | 2, 3 | 349 | |||

Gdpd1
Genomic Map