siRNA Details
| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005582.2 | NP_005573.2 | 2 | 351 | ||
| XM_005248504.4 | XP_005248561.1 | 3 | 360 | |||
| GenBank mRNA | AK313177.1 | - | 2 | 361 | ||
| AL544501.3 | - | 2 | 351 | |||
| BC109069.1 | - | 2 | 309 | 40034168 | ||
| BC109070.1 | - | 2 | 309 | 40034172 | ||
| BP344300.1 | - | 1 | 13 | |||
| D83597.1 | - | 2 | 345 | |||

CD180
Genomic Map