siRNA Details
| Gene Symbol: Uqcrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025641.3 | NP_079917.1 | 3 | 192 | ||
| GenBank mRNA | AK003266.1 | - | 3 | 157 | ||
| AK003539.1 | - | 3 | 135 | |||
| AK011734.1 | - | 554 | ||||
| AK012552.1 | - | 3 | 170 | |||
| AK018712.1 | - | 3 | 170 | |||
| AK054005.1 | - | 3 | 158 | |||
| AK088185.1 | - | 3 | 157 | |||
| BC011388.1 | - | 3 | 136 | 3597862 | ||
| BY138998.1 | - | 3 | 192 | |||
| BY702808.1 | - | 3 | 165 | |||

Uqcrh
Genomic Map