siRNA Details
| Gene Symbol: Crls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024385.1 | NP_001019556.1 | 2 | 569 | ||
| NM_025646.3 | NP_079922.2 | 2 | 256 | |||
| GenBank mRNA | AK002408.1 | - | 2 | 571 | ||
| AK004215.1 | - | 2 | 285 | |||
| AK016165.1 | - | 2 | 257 | |||
| AK017718.1 | - | 2 | 548 | |||
| AK018180.1 | - | 2 | 570 | |||
| BC048702.1 | - | 2 | 550 | 6506241 | ||
| BC086619.1 | - | 2 | 528 | |||

Crls1
Genomic Map