siRNA Details
| Gene Symbol: Crls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024385.1 | NP_001019556.1 | 4, 5 | 821 | ||
| NM_025646.3 | NP_079922.2 | 4, 5 | 508 | |||
| GenBank mRNA | AK002408.1 | - | 4, 5 | 823 | ||
| AK004215.1 | - | 4, 5 | 537 | |||
| AK016165.1 | - | 4, 5 | 509 | |||
| AK017718.1 | - | 3, 4 | 670 | |||
| AK018180.1 | - | 4, 5 | 822 | |||
| BC048702.1 | - | 4, 5 | 802 | 6506241 | ||
| BC086619.1 | - | 4, 5 | 778 | |||

Crls1
Genomic Map