siRNA Details
| Gene Symbol: Fastk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023229.2 | NP_075718.2 | 8 | 1471 | ||
| GenBank mRNA | AF168682.1 | - | 7 | 1366 | ||
| AK002552.1 | - | 8 | 1419 | |||
| AK003940.1 | - | 5 | 714 | |||
| AK038596.1 | - | 8 | 1423 | |||
| AK052667.1 | - | 8 | 1416 | |||
| AK139374.1 | - | 7 | 2191 | |||
| AK151859.1 | - | 8 | 1423 | |||
| AK152369.1 | - | 8 | 1472 | |||
| AK159751.1 | - | 8 | 1423 | |||
| AK164317.1 | - | 1429 | ||||
| AK166508.1 | - | 2016 | ||||
| AK167346.1 | - | 7 | 2183 | |||
| AK186668.1 | - | 1 | 119 | |||
| AK187385.1 | - | 1 | 120 | |||
| BC013547.1 | - | 8 | 1388 | 4218374 | ||

Fastk
Genomic Map