siRNA Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025652.3 | NP_079928.2 | 7 | 980 | ||
| GenBank mRNA | AF391799.1 | - | 7 | 964 | ||
| AK008247.2 | - | 7 | 948 | |||
| AK011845.1 | - | 6 | 1469 | |||
| AK012797.1 | - | 7 | 927 | |||
| AK017234.1 | - | 7 | 874 | |||
| AK143627.1 | - | 7 | 932 | |||
| BC032292.1 | - | 6 | 635 | 5374268 | ||
| BC057966.1 | - | 6 | 1192 | |||
| BC064891.1 | - | 5 | 651 | |||

Gtf3a
Genomic Map