siRNA Details
| Gene Symbol: Trim13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164220.1 | NP_001157692.1 | 3 | 858 | ||
| NM_023233.3 | NP_075722.1 | 3 | 796 | |||
| GenBank mRNA | AK013959.2 | - | 3 | 768 | ||
| AK017850.1 | - | 2 | 753 | |||
| AK162129.1 | - | 3 | 846 | |||
| BC138576.1 | - | 2 | 722 | 8861596 | ||
| BC145915.1 | - | 2 | 722 | 40131218 | ||

Trim13
Genomic Map