siRNA Details
| Gene Symbol: 3110001I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025653.2 | NP_079929.1 | 2 | 1002 | ||
| GenBank mRNA | AK012939.1 | - | 2 | 1003 | ||
| AK013950.1 | - | 2 | 1066 | |||
| AK031567.1 | - | 2 | 1036 | |||
| BC038880.1 | - | 1 | 821 | 1477704 | ||

3110001I22Rik
Genomic Map