siRNA Details
| Gene Symbol: 3110001I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025653.2 | NP_079929.1 | 2 | 765 | ||
| GenBank mRNA | AK012939.1 | - | 2 | 766 | ||
| BC038880.1 | - | 1 | 584 | 1477704 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110001I22Rik| Gene Symbol: 3110001I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025653.2 | NP_079929.1 | 2 | 765 | ||
| GenBank mRNA | AK012939.1 | - | 2 | 766 | ||
| BC038880.1 | - | 1 | 584 | 1477704 | ||