siRNA Details
| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159609.1 | NP_001153081.1 | 3 | 329 | ||
| NM_001159610.1 | NP_001153082.1 | 3 | 380 | |||
| NM_001159612.1 | NP_001153084.1 | 3 | 396 | |||
| NM_025657.3 | NP_079933.2 | 3 | 279 | |||
| GenBank mRNA | AK003037.1 | - | 3 | 186 | ||
| AK003592.1 | - | 3 | 297 | |||
| AK012637.1 | - | 3 | 394 | |||
| AK015684.1 | - | 3 | 330 | |||
| AK033084.1 | - | 1 | 591 | |||
| AK134971.1 | - | 3 | 238 | |||
| AK169249.1 | - | 3 | 303 | |||
| BC034894.1 | - | 3 | 238 | 3328402 | ||
| BU604191.1 | - | 3 | 282 | |||
| BY293050.1 | - | 3 | 255 | |||
| CJ169080.1 | - | 3 | 398 | |||

Lrrc57
Genomic Map