siRNA Details
| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026994.1 | NP_081270.1 | 10 | 1097 | ||
| NM_133679.2 | NP_598440.1 | 11 | 1142 | |||
| GenBank mRNA | AK008846.1 | - | 11 | 915 | ||
| AK017995.1 | - | 10 | 1090 | |||
| AK145827.1 | - | 11 | 966 | |||
| AK170997.1 | - | 3 | 258 | |||
| BC010479.1 | - | 11 | 939 | 3968727 | ||
| BC019387.1 | - | 10 | 906 | 4164847 | ||

Cryzl1
Genomic Map