siRNA Details
| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026994.1 | NP_081270.1 | 5 | 516 | ||
| NM_133679.2 | NP_598440.1 | 6 | 561 | |||
| GenBank mRNA | AK008846.1 | - | 6 | 337 | ||
| AK010433.1 | - | 6 | 365 | |||
| AK017995.1 | - | 5 | 517 | |||
| AK145827.1 | - | 6 | 385 | |||
| BC010479.1 | - | 6 | 358 | 3968727 | ||
| BC019387.1 | - | 5 | 331 | 4164847 | ||
| BY717916.1 | - | 5 | 517 | |||

Cryzl1
Genomic Map