siRNA Details
| Gene Symbol: Gpatch4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110809.2 | NP_001104279.1 | 8 | 1299 | ||
| NM_001286857.1 | NP_001273786.1 | 8 | 1913 | |||
| NM_025663.4 | NP_079939.3 | 9 | 1434 | |||
| GenBank mRNA | AK011617.1 | - | 8 | 1297 | ||
| AK169111.1 | - | 8 | 1288 | |||
| BC019490.1 | - | 10 | 1155 | 4039649 | ||
| BC080676.1 | - | 8 | 1891 | |||
| BU705305.1 | - | 1 | 46 | |||
| CF616874.1 | - | 1 | 136 | |||

Gpatch4
Genomic Map