siRNA Details
| Gene Symbol: Snx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025664.5 | NP_079940.2 | 9 | 1027 | ||
| GenBank mRNA | AK012520.1 | - | 4 | 319 | ||
| AK139802.1 | - | 9 | 1061 | |||
| AK149739.1 | - | 9 | 1041 | |||
| AK149849.1 | - | 9 | 1081 | |||
| AK155830.1 | - | 1062 | ||||
| AK157517.1 | - | 9 | 1043 | |||
| AK170339.1 | - | 1043 | ||||
| BC014814.1 | - | 9 | 1027 | 3257422 | ||

Snx9
Genomic Map