siRNA Details
| Gene Symbol: Snrnp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025665.2 | NP_079941.1 | 5 | 381 | ||
| GenBank mRNA | AK011937.1 | - | 5 | 382 | ||
| AK045231.1 | - | 5 | 394 | |||
| AK160190.1 | - | 5 | 391 | |||
| AK210627.1 | - | 57 | ||||
| BC027564.1 | - | 5 | 382 | 3468252 | ||

Snrnp27
Genomic Map