siRNA Details
| Gene Symbol: Ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025666.2 | NP_079942.1 | 4 | 456 | ||
| GenBank mRNA | AK011968.1 | - | 4 | 327 | ||
| AK017542.1 | - | 4 | 412 | |||
| AK088208.1 | - | 4 | 458 | |||
| AK146308.1 | - | 4 | 457 | |||
| AK149989.1 | - | 4 | 411 | |||
| AK155611.1 | - | 4 | 327 | |||
| AK190790.1 | - | 3 | 190 | |||
| BC058535.1 | - | 3 | 140 | |||
| BC079531.1 | - | 3 | 153 | |||
| BC094323.1 | - | 4 | 463 | |||
| BC094423.1 | - | 4 | 414 | 6812389 | ||

Ubr7
Genomic Map