siRNA Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025670.4 | NP_079946.2 | 6 | 1279 | ||
| GenBank mRNA | AK005157.1 | - | 6 | 1265 | ||
| AK017480.1 | - | 6 | 1262 | |||
| AK031705.1 | - | 6 | 1281 | |||
| AK035791.1 | - | 4 | 2574 | |||
| AK051560.1 | - | 6 | 1277 | |||
| AK149939.1 | - | 6 | 1275 | |||
| AK150219.1 | - | 6 | 1224 | |||
| AK166141.1 | - | 6 | 1272 | |||
| BC004063.1 | - | 6 | 1278 | 3590511 | ||

Cdip1
Genomic Map