siRNA Details
| Gene Symbol: Hiatl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083901.1 | NP_001077370.1 | 9 | 1142 | ||
| NM_133680.3 | NP_598441.3 | 8 | 1239 | |||
| GenBank mRNA | AK146596.1 | - | 9 | 1135 | ||
| BC018395.1 | - | 1 | 27 | |||
| BC033469.1 | - | 8 | 1201 | 4951631 | ||

Hiatl1
Genomic Map