siRNA Details
| Gene Symbol: Hiatl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083901.1 | NP_001077370.1 | 7 | 928 | ||
| NM_133680.3 | NP_598441.3 | 6 | 1025 | |||
| GenBank mRNA | AK146596.1 | - | 7 | 921 | ||
| BC033469.1 | - | 6 | 987 | 4951631 | ||
| CN709077.1 | - | 3 | 354 | |||

Hiatl1
Genomic Map