siRNA Details
| Gene Symbol: Dph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025675.4 | NP_079951.1 | 5 | 535 | ||
| GenBank mRNA | AK006965.1 | - | 5 | 493 | ||
| AK007021.1 | - | 5 | 538 | |||
| AK017595.1 | - | 5 | 491 | |||
| AK036021.1 | - | 5 | 571 | |||
| AK082013.1 | - | 5 | 572 | |||
| AK088160.1 | - | 5 | 537 | |||
| AK145789.1 | - | 5 | 495 | |||
| BC022995.1 | - | 5 | 560 | 5363234 | ||

Dph6
Genomic Map