siRNA Details
| Gene Symbol: Dph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025675.4 | NP_079951.1 | 6, 7 | 636 | ||
| GenBank mRNA | AK006965.1 | - | 6, 7 | 594 | ||
| AK007021.1 | - | 6, 7 | 639 | |||
| AK017595.1 | - | 6, 7 | 592 | |||
| AK036021.1 | - | 6, 7 | 672 | |||
| AK082013.1 | - | 6, 7 | 673 | |||
| AK088160.1 | - | 6, 7 | 638 | |||
| AK145789.1 | - | 6, 7 | 596 | |||

Dph6
Genomic Map