siRNA Details
| Gene Symbol: Mcm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291054.1 | NP_001277983.1 | 9 | 1287 | ||
| NM_025676.4 | NP_079952.2 | 8 | 1203 | |||
| GenBank mRNA | AK017607.1 | - | 9 | 1185 | ||
| AK133858.1 | - | 9 | 1244 | |||
| BC046780.1 | - | 9 | 1234 | 5716136 | ||
| BC052070.1 | - | 8 | 1009 | 5716125 | ||

Mcm8
Genomic Map