siRNA Details
| Gene Symbol: Tsen15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025677.3 | NP_079953.2 | 1, 2 | 224 | ||
| GenBank mRNA | AK004261.1 | - | 1, 2 | 160 | ||
| AK011608.1 | - | 1, 2 | 181 | |||
| AK017650.1 | - | 1, 2 | 225 | |||
| BY065176.1 | - | 1, 2 | 176 | |||

Tsen15
Genomic Map