siRNA Details
| Gene Symbol: Rpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025683.2 | NP_079959.2 | 3 | 251 | ||
| GenBank mRNA | AK007735.1 | - | 3 | 257 | ||
| AK017774.1 | - | 3 | 252 | |||
| AK029088.1 | - | 3 | 250 | |||
| AK088152.1 | - | 3 | 252 | |||
| AK150008.1 | - | 3 | 289 | |||
| AK185217.1 | - | 3 | 245 | |||
| BC019126.1 | - | 3 | 294 | 5008075 | ||
| Z78161.1 | - | 4 | 270 | |||

Rpe
Genomic Map