siRNA Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000239.2 | NP_000230.1 | 2 | 270 | ||
| GenBank mRNA | AK130149.1 | - | 2 | 275 | ||
| AK311779.1 | - | 2 | 243 | |||
| BC004147.2 | - | 2 | 234 | 2959387 | ||
| FJ795023.1 | - | 2 | 237 | |||
| J03801.1 | - | 2 | 240 | |||
| KJ841928.1 | - | 2 | 215 | |||
| M19045.1 | - | 2 | 228 | |||
| M21119.1 | - | 2 | 228 | |||
| U25677.1 | - | 2 | 215 | |||

LYZ
Genomic Map