siRNA Details
| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175111.3 | NP_780320.2 | 8 | 1444 | ||
| GenBank mRNA | AK017731.1 | - | 8 | 1399 | ||
| AK028824.1 | - | 9 | 1505 | |||
| AK076189.1 | - | 8 | 1411 | |||
| AK089941.1 | - | 8 | 1460 | |||

Hspbap1
Genomic Map