siRNA Details
| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025697.3 | NP_079973.1 | 4 | 673 | ||
| GenBank mRNA | AK018044.1 | - | 4 | 551 | ||
| AK018153.1 | - | 4 | 674 | |||
| AK018290.1 | - | 4 | 523 | |||
| AK137685.1 | - | 522 | ||||
| AK167306.1 | - | 4 | 515 | |||
| AK169059.1 | - | 4 | 522 | |||
| BC037015.1 | - | 4 | 507 | 5344409 | ||

Spryd7
Genomic Map