siRNA Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175112.5 | NP_780321.1 | 11 | 1119 | ||
| GenBank mRNA | AK014325.1 | - | 11 | 1075 | ||
| AK017746.1 | - | 11 | 1040 | |||
| AK035278.1 | - | 11 | 1065 | |||
| AK079383.1 | - | 11 | 1100 | |||
| AK146058.1 | - | 11 | 1084 | |||
| AK156681.1 | - | 11 | 1084 | |||
| AK177228.1 | - | 1 | 35 | |||
| AK180304.1 | - | 2 | 106 | |||
| AK217457.1 | - | 2 | 106 | |||
| BC059051.1 | - | 11 | 1041 | 6818771 | ||
| BC060072.1 | - | 9 | 3580 | 5718934 | ||

Rae1
Genomic Map