siRNA Details
| Gene Symbol: Oser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025699.2 | NP_079975.2 | 3 | 251 | ||
| GenBank mRNA | AK006969.1 | - | 3 | 294 | ||
| AK009708.1 | - | 3 | 267 | |||
| AK014323.1 | - | 3 | 270 | |||
| AK088622.1 | - | 3 | 270 | |||
| AK139692.1 | - | 423 | ||||
| AK153499.1 | - | 3 | 288 | |||
| BC005579.1 | - | 3 | 251 | 3968876 | ||

Oser1
Genomic Map