siRNA Details
| Gene Symbol: Trappc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025701.4 | NP_079977.3 | 2 | 425 | ||
| GenBank mRNA | AK003633.1 | - | 2 | 393 | ||
| AK007989.1 | - | 2 | 392 | |||
| AK076212.1 | - | 2 | 426 | |||
| AK078451.1 | - | 2 | 391 | |||
| AK171833.1 | - | 1 | 2805 | |||
| BC028946.1 | - | 1 | 885 | |||
| BC049179.1 | - | 2 | 383 | 6313540 | ||

Trappc5
Genomic Map