siRNA Details
| Gene Symbol: Snx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025712.4 | NP_079988.3 | 10, 11 | 1047 | ||
| GenBank mRNA | AK014536.1 | - | 10, 11 | 1049 | ||
| AK034196.1 | - | 10, 11 | 1049 | |||
| AK035226.1 | - | 10, 11 | 1048 | |||
| BC061007.1 | - | 10, 11 | 1043 | 30314775 | ||

Snx31
Genomic Map