siRNA Details
| Gene Symbol: Rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025717.3 | NP_079993.2 | 3 | 753 | ||
| GenBank mRNA | AK006447.1 | - | 3 | 736 | ||
| AK014823.1 | - | 3 | 673 | |||
| AK142116.1 | - | 3 | 709 | |||
| AK160796.1 | - | 3 | 1148 | |||
| AK181241.1 | - | 1 | 160 | |||
| AK182975.1 | - | 1 | 160 | |||
| BC019488.1 | - | 3 | 658 | 4457936 | ||

Rbm4b
Genomic Map